Order Entry
Denmark
ContactUsLinkComponent
Red hydrogen peroxide assay kit
Red hydrogen peroxide assay kit
Catalog # ENZOENZ51004
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Red hydrogen peroxide assay kit
Catalog # ENZOENZ51004
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Red hydrogen peroxide assay kit
  • Seq:
    02
  • Storage conditions:
    –20 °C (Long term)
  • Application:
    Fluorescence microscopy, HTS
  • Size:
    1 kit

Specifications

About this item

Red Hydrogen Peroxide Assay Kit provides a simple absorbance-based or fluorometric assay to detect and quantify hydrogen peroxide or peroxidase activity in biochemical assays, cell extracts and in live cells. It can also be used to detect a variety of oxidase activities through enzyme-coupled reactions. The kit is an optimized ‘mix and read’ assay that is suitable for both 96- and 348-well formats and is compatible with HTS liquid handling.

  • Homogenous assay format, fully compatible with HTS automation
  • Detect as little as 10 picomoles of hydrogen peroxide
  • Mix and read reagents, with minimal hands-on time
Delivery: Kit includes Red Peroxidase Substrate (1 Vial), Horseradish Peroxidase (HRP) (20 units), Hydrogen Peroxide (H2O2) Stock Solution (200 µl), DMSO (1 ml), Assay Buffer (100 ml).