Order Entry
Denmark
ContactUsLinkComponent
APO-BRDU™ kit, TUNEL Assay
APO-BRDU™ kit, TUNEL Assay
Catalog # ENZOALX850042KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
APO-BRDU™ kit, TUNEL Assay
Catalog # ENZOALX850042KI01
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    APO-BRDU™ kit
  • Seq:
    02
  • Size:
    1 kit
Specifications
About this item
Kit for measuring apoptosis with high sensitivity by Dual Color Flow Cytometry in mammalian cells. Br-dUTP is more readily incorporated into the genome of apoptotic cells than are larger ligands like fluorescein, biotin, etc. This greater incorporation coupled with the use of a fluorescein labelled anti-BrdU monoclonal antibody (Prod. No. ALX-804-197L) gives rise to a stronger signal in flow cytometry.
Ordering information: Quantity: 50 assays. Contents: Contains all the reagents necessary, including positive and negative control cells.