IgE Antibody
ENZOBPDDIAHE11
New Product
- Host:
- Isotype:IgE
- Reactivity:
- Seq:03
- Environmentally Preferable:
- Form:Liquid
- Purity:≥98% (SDS-PAGE)
- Storage buffer:0,01 M phosphate buffer, pH 7,4, containing 0,15 M sodium chloride
- Pk:1 mg
Purified human IgE with kappa light chains produced in vitro from a monoclonal hybridoma.
Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype: IgE
Reactivity: