Anti-ACTB Rabbit Polyclonal Antibody
Catalog #
Product Details & Documents
Actin is one of the major proteins of the cytoskeleton. It regulates contractile potential in muscle tissues and helps to control the shape as well as the motility of non-muscle cells. Actin is expressed in all eukaryotic cells. In higher eukaryotes, there are six isoforms of actin.
- For Immunocytochemistry, Western Blot
Recognizes human, mouse and rat beta-Actin. Detects a band of ~43kDa by Western blot.
Type: Primary
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human



