Order Entry
Denmark
ContactUsLinkComponent
proBDNF Rapid ELISA Kit: Human, Mouse, Rat (1 Plate)
proBDNF Rapid ELISA Kit: Human, Mouse, Rat (1 Plate)
Catalog # BSENBEK-2237-1P
Supplier:  Biosensis
CAS Number:  
undefined
proBDNF Rapid ELISA Kit: Human, Mouse, Rat (1 Plate)
Catalog # BSENBEK-2237-1P
Supplier:  Biosensis
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    proBDNF
  • Description:
    proBDNF Rapid ELISA Kit
  • Storage conditions:
    Do not freeze.
  • Time to Results:
    <4 hrs
  • Detection range:
    15.6 - 1000 pg/ml
  • Storage temperature:
    2...8 °C
  • Detection limit:
    Typical for proBDNF is 10 pg/mL determined as 150% of the blank value.
  • Tests per kit:
    96 tests (1 microplate)

Specifications

About this item

The Biosensis proBDNF (Brain-derived neurotrophic factor, BDNF, Abrineurin) RapidTM enzyme-linked immunosorbent assay (ELISA) Kit is a sandwich ELISA that allows the specific, fast and reliable quantification of proBDNF in less than 4 hours in cell culture supernatants, human serum and EDTA-plasma only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application, in particular human blood samples.

This ELISA kit consists of a pre-coated monoclonal anti-proBDNF capture antibody, a biotinylated anti-matureBDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of proBDNF present in samples and protein standards. A proBDNF positive control (QC sample) is provided to assure consistent assay performance.

This proBDNF ELISA kit employs a recombinant, cleavage-resistant human proBDNF standard produced in mammalian cells by Biosensis and validated against externally available proBDNF proteins.

Note that accurate proBDNF quantification in human serum and EDTA-plasma requires the addition of Heterophilic Antibody Blocker BL-004-500 provided in the kit, and available for purchase separately.

This ELISA kit has not been tested for other applications. It has been configured for research use only and is not to be used for diagnostic or clinical procedures. The ELISA kit box contains 2× 96 pre-coated strip plate, protein standards, QC sample, detection reagents, heterophilic antibody blocker, wash and sample buffers and detailed protocols.