Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, (1S)-(−)-Verbenone
Supelco® Organic Reference Standard, (1S)-(−)-Verbenone
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, (1S)-(−)-Verbenone
Supplier:  Merck

Specifications

  • CAS No.:
    1196-01-6
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00065445
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (1S,5S)-2-Pinen-4-one; (1S,5S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-one
  • Molecular weight:
    150,22
  • Density:
    0,975 g/ml at 20 °C (lit.)
  • Flash point:
    85 °C - Closed cup
  • Boiling point:
    227 - 228 °C (lit.)

Specifications

Frequently Bought Together