
Citalopram hydrobromide (by HPLC)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59729-32-7
:
Formula:C₂₀H₂₁FN₂O
MDL Number:MFCD02101306
Melting Pt:185…188 °C
:405.31 g/mol
:GHS07
Storage Temperature:Køleskab
Specification Test Results
- Appearance:White to almost white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105°C, 4 h)
- Solubility:(1 % in ethanol) Clear colorless
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together