Hydrochloric acid 0.1 M (0.1 N) volumetric solution
:7647-01-0
:
:HCl
:MFCD00011324
:8,III
:36.46 g/mol
:1789
- Molarity:0,1 M
- Appearence:Clear colourless liquid
- pH :1,2
- Titer at 20 °C:0,999 – 1,001
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Qualitative filter papers, Grade 2555 ½, Whatman™
: Whatman products (Cytiva)
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection