0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Paraformaldehyde
Paraformaldehyde 4% in PBS 91-93%
Paraformaldehyde 4% in PBS 91-93%
ALFAJ19943.K2
:
Thermo Fisher Scientific
:
Paraformaldehyde 4% in PBS 91-93%
ALFAJ19943.K2
:
Thermo Fisher Scientific
:
Pk:
1 L
Seq:
5037
Form:
Colourless, clear solution
Assay (unspecified) (formaldehyde):
91 − 93%
pH (25 °C):
7,0 − 7,6
Concentration:
1XPBS (0,01M)
:
:
30525-89-4
:
(CH₂O)n
:
MFCD00133991