Anti-COX1/Cyclooxygenase 1 Polyclonal Antibody
Catalog # BSBTPB9002
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:COX1/Cyclooxygenase 1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for COX 1/PTGS1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. COX 1/PTGS1 information: Molecular Weight: 68686 MW; Subcellular Localization: Microsome membrane; Peripheral membrane protein. Endoplasmic reticulum membrane; Peripheral membrane protein.
Type: Primary
Antigen: COX1/Cyclooxygenase 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat