Order Entry
Denmark
ContactUsLinkComponent
 
Pyrantel pamoate 98%
 
undefined
Pyrantel pamoate 98%

 

  • Appearance:
    Yellow to brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 % (60°C, 3 h) (vacuum)
  • Sulfated ash:
    ≤0.5 %
  • Titration with HClO4:
    97 to 103 % (on dry substance)

 

Frequently Bought Together