Order Entry
Denmark
ContactUsLinkComponent
Caffeine citrate
Caffeine citrate
Certificates
undefined
Caffeine citrate
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 %
  • Titration with HClO4:
    48 to 52 % (Caffeine)
  • Titration with TBAH:
    48 to 52 % (Citric acid)