Order Entry
Denmark
ContactUsLinkComponent
(1-Methyl-1H-pyrazol-5-yl)methylamine 97%
undefined
(1-Methyl-1H-pyrazol-5-yl)methylamine 97%
  • Appearance:
    Clear colorless to yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1.5 %
  • GC:
    ≥96.0 %
Frequently Bought Together