Tin(IV) oxide 99%, pure
Leverandør: Thermo Fisher Scientific
Certifikater
Om denne vare
CAS nummer:18282-10-5
:
Formel:O₂Sn
MDL nummer:MFCD00011244
:150.71 g/mol
Tæthed:6.95 g/cm³
Opbevaringstemperatur:Ambient
Specifikationstestresultater
- Appearance:White to light grey Powder
- Purity:≥98.5 % (metals basis)
Specifikationstestresultater
Frequently Bought Together