


Petroleum spirit, 40…60 °C, Laboratory Reagent
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64742-49-0
:8032-32-4
MDL Number:MFCD00081849
Boiling Pt:40…60 °C (1.013 hPa)
Density:0.65 g/cm³ (20 °C)
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:3295
ADR:3,II
:GHS07
:GHS08
:GHS02
Specification Test Results
- Boiling range:40 - 60 °C
- Water (Karl Fischer):Max. 0.02 %
- Non-volatile matter:Max. 0.002 %
- Aromatic substances (as C₆H₆):Max. 0.01 %
Specification Test Results
Frequently Bought Together