Organic reference standard, Geosmin, Dr. Ehrenstorfer
Catalog # EHERLA14005000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:1 mL
- Standards:Reference standard
- CAS No.:16423-19-1
- Description:(±)-Geosmin 10 µg/ml in methanol
- Formula:C₁₂H₂₂O
- Description:(±)-Geosmin
Specifications
About this item
This standard solution is designed for calibration or recalibration of chromatographic systems for the determination of the specified chemical compounds concerning identity and qualification. It can be also used as reference material for interlaboratory studies to validate analytical procedures.
A serial numbered certificate is provided from an ISO 17025 calibration to indicate the instrument standard.
Certifications: Accredited to ISO/IEC 17025.