ASTM® precision thermometers, low hazard
Supplier: Amarell
Certificates
About this item
Low hazard precision thermometers are used for special applications and suitable for government calibration.
- Solid stem and white backed
- Round or prismatic with round top finish
- Durable pigment
- Non-wetting because of capillary tube specially coated inside
Certifications: ASTM standard E2251–07 (Standard specification for liquid-in-glass ASTM thermometers with low-hazard precision liquids).
Packaging: Packaged in transparent square plastic case.
Frequently Bought Together