Calbiochem® human IgM (from plasma)
1 mg
Catalog # 401799-1
Supplier: Merck Millipore (Oncogene)
Calbiochem® human IgM (from plasma)
Catalog # 401799-1
Supplier: Merck Millipore (Oncogene)
CAS Number:
Jump to:
Product Details & Documents
Prepared from plasma shown to be nonreactive for HBsAg and for antibodies to HCV, HBc, HIV-1, and HIV-2.
Specifications
Specifications
- Pk:1 mg
- Cat. No.:401799-1
- Protein/peptide name:IgM
- Storage conditions:–20 °C
- Formulation:In 200 mM NaCl, 50 mM Tris, pH 8,0
- Purity:>95% by SDS-PAGE
- Species:Human
- Source:Plasma



