Filter tips, Qualitix®
These tips are made of 100% pure virgin, medical grade PP, fitting not only Socorex® but all major micropipette brands. They include a hydrophobic HDPE filter with 10 µ pore size to block any contaminant.
- Superior quality, autoclavable
- Efficient protection against aerosols
- Lot by lot consistency
- Certified free of nuclease, endotoxin and ATP
- Reduced packaging size, less bench space requirement
- Broad compatibility with other micropipette brands
Beveled tips: Special shape eliminates liquid drop sticking at tip end.
Fine pointed tips: Have a thin, soft end enabling smooth liquid deposit.
Frequently Bought Together