Order Entry
Denmark
Orders LinkContactUsLinkComponent
ISE standard solutions
 :   WTW
ISE standard solutions
 :  WTW
ISE standard solutions
ISE standard solutions
These standard solutions for calibration contain 10 g of the ion to be measured per 10 liters.
Product Family Options
  • Description
    Pk
  • Standard solution chloride (ES/Cl)
    1 L
    1 * 1 L
    1 * 1 L - 1.670,00 kr
    1.670,00 kr
    1 * 1 L
       1.670,00 kr
  • Standard solution fluoride (ES/F)
    1 L
    1 * 1 L
    1 * 1 L - 1.670,00 kr
    1.670,00 kr
    1 * 1 L
       1.670,00 kr
  • Standard solution calcium ions (ES/Ca)
    1 L
    1 * 1 L
    1 * 1 L - 1.670,00 kr
    1.670,00 kr
    1 * 1 L
       1.670,00 kr
  • Standard solution potassium ions (ES/K)
    1 L
    1 * 1 L
    1 * 1 L - 1.670,00 kr
    1.670,00 kr
    1 * 1 L
       1.670,00 kr
  • Standard solution nitrate (ES/NO₃)
    1 L
    1 * 1 L
    1 * 1 L - 1.670,00 kr
    1.670,00 kr
    1 * 1 L
       1.670,00 kr
  • Standard solution sodium ions (ES/Na)
    1 L
    1 * 1 L
    1 * 1 L - 1.670,00 kr
    1.670,00 kr
    1 * 1 L
       1.670,00 kr