Organic reference standard, Ursodeoxycholic acid, Ph.Eur., RS
Catalog # EDQMU0800000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:128-13-2
- Description:Ursodeoxycholic acid
- Formula:C₂₄H₄₀O₄
- Storage temperature:Ambient
- Description:Ursodeoxycholic acid
Specifications
About this item
- In Monographs 1189 and 1275
- Solubility in water: less than 0.02 g/l at 20°C