Organic reference standard, Triamcinolone acetonide, Ph.Eur., RS
EDQMT1900000
: EDQM
:
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:76-25-5
- Description:Triamcinolone acetonide
- Formula:C₂₄H₃₁FO₆
- Storage temperature:Ambient
- Description:Triamcinolone acetonide
- In Monographs 0533, 0867, and 1075