Organic reference standard, Pentamidine diisetionate, Ph.Eur., RS
Catalog # EDQMP0405000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:30 mg
- Standards:Reference standard
- CAS No.:140-64-7
- Description:Pentamidine diisetionate
- Formula:C₂₃H₃₆N₄O₁₀S₂
- Storage temperature:Ambient
- Description:Pentamidine diisetionate
Specifications
About this item
- In Monograph 1137 and 1436
- Solubility in water: 100 to 1000 g/l