Organic reference standards, Levamisole hydrochloride, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:16595-80-5
- Formula:C₁₁H₁₃ClN₂S
- Storage temperature:Ambient
- Description:Levamisole hydrochloride for system suitability
Specifications
Frequently Bought Together