Organic reference standards, Ketoprofen, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
- In Monograph 0922
Specifications
- CAS No.:66067-44-5
- Formula:C₁₅H₁₂O₂
- Storage temperature:Ambient
- Description:Ketoprofen impurity A
Specifications
Frequently Bought Together