Organic reference standard, Itraconazole, Ph.Eur., RS
EDQMI7000000
: EDQM
:
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:84625-61-6
- Description:Itraconazole
- Formula:C₃₅H₃₈Cl₂N₈O₄
- Storage temperature:Ambient
- Description:Itraconazole
- In Monograph 1335
- Solubility in water: less than 0,1 g/l