Organic reference standards, Ibuprofen, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
- In Monograph 0721
Specifications
- CAS No.:15687-27-1
- Formula:C₁₃H₁₈O₂
- Storage temperature:Ambient
- Description:Ibuprofen
Specifications
Frequently Bought Together