Organic reference standard, Enalapril, for system suitability, Ph.Eur., RS
Catalog # EDQME0110000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:10 mg
- Standards:Reference standard
- CAS No.:76095-16-4
- Description:Enalapril, for system suitability
- Formula:C₂₄H₃₂N₂O₉
- Storage temperature:Ambient
- Description:Enalapril, for system suitability
Specifications
About this item
A long-acting angiotensin-converting enzyme inhibitor.
- In Monograph 1420
- Solubility in water: 10 - 33,3 g/l
- Soluble in/miscible with methanol