Organic reference standard, Chlorphenamine maleate, Ph.Eur., RS
Catalog # EDQMC1800000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:20 mg
- Standards:Reference standard
- CAS No.:113-92-8
- Description:Chlorphenamine maleate
- Formula:C₂₀H₂₃ClN₂O₄
- Storage temperature:Ambient
- Description:Chlorphenamine maleate
Specifications
About this item
- In Monographs 0386,0977,1084, and 1196