Organic reference standard, Cefradine CRS, Ph.Eur., RS
Catalog # EDQMC0690000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:300 mg
- Standards:Reference standard
- CAS No.:38821-53-3
- Description:Cefradine CRS
- Formula:C₁₆H₁₉N₃O₄S
- Storage temperature:Ambient
- Description:Cefradine CRS
Specifications
About this item
Filled under inert atmosphere and controlled humidity.
- Partial solubility in water