Ipratropium Bromide, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Catalog No:
- 1347507.
- 1347518.
- 1347529.
- 1347530.
- Description:
- Ipratropium Bromide
- Ipratropium Bromide Related Compound A
- Ipratropium Bromide Related Compound B
- Ipratropium Bromide Related Compound C
- Synonym(s):
- (1R,3r,5S,8r)-3-Hydroxy-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane, bromide
- (1R,3r,5S,8s)-3-[[(2RS)-3-hydroxy-2-phenylpropanoyl]oxy]-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane, bromide
- (2-RS)-3-hydroxy-2-phenylpropanoic acid
- CAS No.:
- 66985-17-9
- 58005-18-8
- 58073-59-9
- 552-63-6
- Formula:
- C₂₀H₃₂BrNO₄
- C₁₁H₂₂BrNO
- C₂₀H₃₀BrNO₃
- C₉H₁₀O₃
- UN No.:
- 1544
Product Family Options
- DescriptionStandardsPack typePk
- Ipratropium BromideUSP reference standardVial200 mg1 * 200 mg3.320,00 kr
Specifications
Cat. No.1347507.DescriptionIpratropium BromideSynonym(s)CAS No.66985-17-9FormulaC₂₀H₃₂BrNO₄UN No. - Ipratropium Bromide Related Compound AUSP reference standard10 mg1 * 10 mg9.540,00 kr
Specifications
Cat. No.1347518.DescriptionIpratropium Bromide Related Compound ASynonym(s)(1R,3r,5S,8r)-3-Hydroxy-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane, bromideCAS No.58005-18-8FormulaC₁₁H₂₂BrNOUN No.1544 - Ipratropium Bromide Related Compound BUSP reference standardVial10 mg1 * 10 mg9.540,00 kr
Specifications
Cat. No.1347529.DescriptionIpratropium Bromide Related Compound BSynonym(s)(1R,3r,5S,8s)-3-[[(2RS)-3-hydroxy-2-phenylpropanoyl]oxy]-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane, bromideCAS No.58073-59-9FormulaC₂₀H₃₀BrNO₃UN No. - Ipratropium Bromide Related Compound CUSP reference standardVial15 mg1 * 15 mg9.730,00 kr
Specifications
Cat. No.1347530.DescriptionIpratropium Bromide Related Compound CSynonym(s)(2-RS)-3-hydroxy-2-phenylpropanoic acidCAS No.552-63-6FormulaC₉H₁₀O₃UN No.



