Order Entry
Denmark
ContactUsLinkComponent
Recombinant NAPRTase B. Subtilis (from E. coli)
Recombinant NAPRTase B. Subtilis (from E. coli)
Catalog # PRSI90-362
Supplier:  ProSci Inc.
CAS Number:  
Recombinant NAPRTase B. Subtilis (from E. coli)
Catalog # PRSI90-362
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at –80 °C.
  • Enzyme name:
    NAPRTase B. Subtilis
  • Enzyme activity:
    ~0,8 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1 umol nicotinic acid mononucleotide (NaMN) per min.
  • Purity:
    >98% (SDS-PAGE)
  • Formulation:
    Liquid, in 20 mM TRIS-HCl, pH 7,8 containing 50 mM sodium chloride.

Specifications

About this item

Nicotinate Phosphoribosyltransferase (NAPRTase) is involved in the biological processes of pyridine nucleotide biosynthesis and nicotinate nucleotide salvage and functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN). It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.

Fusion-Tag: Tag Free

This recombinant proteins is for research use only.