Order Entry
Denmark
ContactUsLinkComponent
 
Mouse recombinant CNDP dipeptidase 2
Catalog # PRSI92-416
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant CNDP dipeptidase 2
Catalog # PRSI92-416
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    CNDP dipeptidase 2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    53.8 kDa
  • Sequence:
    Met1-Asn475
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 8,0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Mouse cytosolic non-specific dipeptidase(CNDP2) is a cytoplasm protein which belongs to the peptidase M20A family. CNDP2 has 2 Isoform: Isoform 1 is ubiquitously expressed with higher levels in kidney and liver (at protein level). Isoform 2 is expressed in fetal tissues, it is only expressed in adult liver and placental tissues. CNDP2 hydrolyses a variety of dipeptides including L-carnosine and has a strong preference for Cys-Gly. It is a cytosolic enzyme that can hydrolyse carnosine to yield l-histidine and beta-alanine. CNDP2 is highly expressed in the histaminergic neurons in the tuberomammillary nucleus, implying that it may supply histidine to histaminergic neurons for histamine synthesis. It may play a role as tumour suppressor in hepatocellular carcinoma (HCC) cells.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.