Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Sedoheptulokinase (from cells)
Human recombinant Sedoheptulokinase (from cells)
Catalog # PRSI92-395
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Sedoheptulokinase (from cells)
Catalog # PRSI92-395
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Sedoheptulokinase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    52.5 kD
  • Sequence:
    Met1-Ser478
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Sedoheptulokinase (SHPK) belongs to the FGGY kinase family, and is mainly located in cytoplasm. SHPK is strongly expressed in liver, kidney and pancreas. It is expressed at lower levels in placenta and heart, and very weakly expressed in lung and brain. SHPK catalyses the chemical reaction: ATP + sedoheptulose = ADP + sedoheptulose 7-phosphatecan, It can transform sedoheptulose to sedoheptulose 7-phosphate in the condition of ATP, and acts as a modulator of macrophage activation through control of glucose metabolism. In addition, It also can be down-regulated by LPS.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.