Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Methylmalonyl-CoA epimerase (from cells)
Human recombinant Methylmalonyl-CoA epimerase (from cells)
Catalog # PRSI92-387
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Methylmalonyl-CoA epimerase (from cells)
Catalog # PRSI92-387
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Methylmalonyl-CoA epimerase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16 kD
  • Sequence:
    Gln37-Ala176
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM DTT, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Methylmalonyl-CoA epimerase, mitochondrial(MCEE)is an enzyme which belongs to the glyoxalase I family. It converts (S)-methylmalonyl-CoA to the (R) form, catalyses the following chemical reaction: (R)-methylmalonyl-CoA (S)-methylmalonyl-CoA. It plays an important role in the catabolism of fatty acids with odd-length carbon chains. This protein deficiency is an autosomal recessive inborn error of AA metabolism, involving valine, threonine, isoleucine and methionine. This organic aciduria can appear in the neonatal period with life-threatening metabolic acidosis, hyperammonemia, feeding difficulties, pancytopenia and coma.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.