Order Entry
Denmark
ContactUsLinkComponent
Human recombinant 6-Phosphogluconate Dehydrogenase (from cells)
Human recombinant 6-Phosphogluconate Dehydrogenase (from cells)
Catalog # PRSI92-379
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant 6-Phosphogluconate Dehydrogenase (from cells)
Catalog # PRSI92-379
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    6-Phosphogluconate Dehydrogenase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    54.2 kD
  • Sequence:
    Met1-Ala483
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

6-phosphogluconate dehydrogenase(PGD) is a cytoplasm-located protein, and belongs to the 6-phosphogluconate dehydrogenase family. 6PGD is the second dehydrogenase in the pentose phosphate shunt. It catalyses the oxidative decarboxylation of 6-phosphogluconate to ribulose 5-phosphate and CO₂, with concomitant reduction of NADP to NADPH. Mutations within the gene coding this enzyme result in 6-phosphogluconate dehydrogenase deficiency, an autosomal hereditary disease effecting the red blood cells.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.