Human recombinant ribonuclease T2 (from cells)
Catalog # PRSI92-334
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Ribonuclease T2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:28.2 kD
- Sequence:Asp25-His256
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHcl, 150 mM NaCl, 20% glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
RNASET2, also known as RNASE6PL, is short for bonuclease T2. It is a 256 aa. protein which belongs to the RNase T2 family. RNASET2 is a secreted protein, and is higher expressed in the temporal lobe and fetal brain. This protein can be inhibited by Zn₂+ and Cu₂+. It has ribonuclease activity, with higher activity at acidic pH and is probably involved in lysosomal degradation of ribosomal RNA. It also plays a role in cellular RNA catabolism.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.