Human recombinant magnesium-dependent phosphatase 1 (from cells)
Catalog # PRSI92-327
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Magnesium-dependent phosphatase 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:21.1 kD
- Sequence:Met1-Ala176
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
MDP1, which is short for nesium-dependent phosphatase 1, is a 176 aa. protein. It has 3 forms by different alternative splicing. This protein belongs to the HAD-like hydrolase superfamily, and usually takes its function with Magnesium. It can be inhibited by vanadate and zinc, and slightly by calcium. MDP1 is a Magnesium-dependent phosphatase which may act as a tyrosine phosphatase.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.