Mouse recombinant Transglutaminase 2
Catalog # PRSI92-325
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Species:Mouse
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Transglutaminase 2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:78 kD
- Sequence:Ala2-Ala686
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 20% glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Biological assays
Specifications
About this item
Protein-glutamine gamma-glutamyltransferase 2 (TGM2) is a 78-kDa, calcium dependent enzyme,It belongs to the transglutaminase superfamily and transglutaminase family. The protein encoded by this TGM2 gene acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. TGM2 is the autoantigen implicated in celiac disease. Two transcript variants encoding different isoforms have been found for this gene.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.