Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Acylphosphate phosphohydrolase 1 (from E. coli)
Human recombinant Acylphosphate phosphohydrolase 1 (from E. coli)
Catalog # PRSI92-308
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Acylphosphate phosphohydrolase 1 (from E. coli)
Catalog # PRSI92-308
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Acylphosphate phosphohydrolase 1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    13.42 kD
  • Sequence:
    Met1-Lys99
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris,100 mM NaCl, 20% glycerol,1 mM DTT,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

ACYP1, also known as Acylphosphatase-1, Acylphosphatase, erythrocyte isozyme, Acylphosphatase, organ-common type isozyme, Acylphosphate phosphohydrolase 1 and ACYPE, is a small cytosolic enzyme which catalyses the hydrolysis of the carboxyl-phosphate bond of acylphosphates. ACYP1 is a protein which belongs to the acylphosphatase family and contains 1 fibrinogen C-terminal domain. Two isoenzymes have been isolated, called muscle acylphosphatase and erythrocyte acylphosphatase, on the basis of their tissue localisation. This gene encodes the erythrocyte acylphosphatase isoenzyme. Alternatively spliced transcript variants that encode different proteins were identified through data analysis. Recombinant human ACYP1 protein was expressed in E. coli fused with HIS-tag at N-terminus.

Fusion-Tag: N-6 His tag.

This recombinant protein can be used for biological assays. For research use only.