Order Entry
Denmark
ContactUsLinkComponent
Human recombinant sulphotransferase 2A1 (from E. coli)
Human recombinant sulphotransferase 2A1 (from E. coli)
Catalog # PRSI92-133
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant sulphotransferase 2A1 (from E. coli)
Catalog # PRSI92-133
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Sulfotransferase 2A1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    35.2 kD
  • Sequence:
    Ser2-Glu285
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 100 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Bile Salt Sulfotransferase (SULT2A1( is a cytosolic enzyme that belongs to the Sulfotransferase 1 family. SULT2A1 is primarily expressed in the liver and adrenal tissues, and to a lesser extent in the kidney. SULT2A1 utilises 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor, and it catalyse the sulfonation of steroids and bile acids in the liver and adrenal glands. SULT2A1 may have a role in the inherited adrenal androgen excess.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.