Order Entry
Denmark
ContactUsLinkComponent
Human recombinant sulphotransferase 1B1 (from E. coli)
Human recombinant sulphotransferase 1B1 (from E. coli)
Catalog # PRSI92-131
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant sulphotransferase 1B1 (from E. coli)
Catalog # PRSI92-131
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Sulfotransferase 1B1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    36.3 kD
  • Sequence:
    Met1-Ile296
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 0.1 M NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Sulfotransferase Family Cytosolic 1B Member 1 (SULT1B1) is a cytosolic enzyme that belongs to the Sulfotransferase 1 family. Human SULT1B1 is a 296 amino acid protein that is highly expressed in the liver, peripheral blood leukocytes, colon, small intestine, and spleen. SULT1B1 utilises 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor, and it can catalyse the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.