Order Entry
Denmark
ContactUsLinkComponent
Human recombinant uridine phosphorylase 1 (from E. coli)
Human recombinant uridine phosphorylase 1 (from E. coli)
  PRSI92-122
 :  
Human recombinant uridine phosphorylase 1 (from E. coli)
  PRSI92-122
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Uridine Phosphorylase 1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21.3 kD
  • Sequence:
    Met1-Ala173
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 200 mM NaCl, 1 mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Uridinephosphorylase 1 (UPP1) is a member of the family of pentosyltransferase. UPP1 catalyses the reversible phosphorolysis of uridine to uracil. The expression levels and the enzymatic activity of UPP1 are higher in human solid tumors than in adjacent normal tissues. The high level of UPP1 expression in some tumors makes it a potential prognosticfactor for some cancers, such as oral squamous cell carcinoma. UPP1 is important for the homeostatic regulation of intracellular and plasma uridine concentratios. UPP1 plays an important role in the pyrimidine salvage pathway through its catalysis of the reversible phosphorolysis of uridine to uracil.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.