Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant Deoxycytidine Kinase (from E. coli)
Human Recombinant Deoxycytidine Kinase (from E. coli)
Catalog # PRSI92-064
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant Deoxycytidine Kinase (from E. coli)
Catalog # PRSI92-064
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His, T7 tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Deoxycytidine Kinase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    34 kD
  • Sequence:
    Met1-Leu260
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Deoxycytidine Kinase (DCK) is a member of the DCK/DGK family. DCK exists as a homodimer and is localised to the nucleus. DCK is required for the phosphorylation of the deoxyribonucleosides deoxycytidine (dC), deoxyguanosine (dG), and deoxyadenosine (dA). DCK has broad substrate specificity, and does not display selectivity based on the chirality of the substrate. In addition, DCK is also an essential enzyme for the phosphorylation of numerous nucleoside analogs widely employed as antiviral and chemotherapeutic agents. DCK is clinically important because of its relationship to drug resistance and sensitivity.

Fusion-Tag: N-6 His, T7 tag

This recombinant protein can be used for biological assays. For research use only.