Order Entry
Denmark
ContactUsLinkComponent
Human recombinant sulphotransferase 1A2 (from E. coli)
Human recombinant sulphotransferase 1A2 (from E. coli)
  PRSI92-054
 :  
Human recombinant sulphotransferase 1A2 (from E. coli)
  PRSI92-054
 :  

 

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Sulfotransferase 1A2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    36.4 kD
  • Sequence:
    Met1-Leu295
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM Tris, 100 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Sulfotransferase 1A2 (SULT1A2) is a member of the Sulfotransferase 1 family. Sulfotransferase enzymes catalyse the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. SULT1A2 is a cytoplasmic protein and exists as a homodimer. SULT1A2 mediates the metabolic activation of carcinogenic N-hydroxyarylamines to DNA binding products and might thus participate as a modulating factor of cancer risk.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.