Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant Kidney-Type Arginase (from E. coli)
Human Recombinant Kidney-Type Arginase (from E. coli)
Catalog # PRSI92-034
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant Kidney-Type Arginase (from E. coli)
Catalog # PRSI92-034
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Kidney-Type Arginase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    40.74 kD
  • Sequence:
    Met1-Ile354
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Kidney-Type Arginase (ARG2) is a member of the arginase family. Arginase is a manganese-containing enzyme which catalyses the hydrolysis of arginine to ornithine and urea. ARG2 is highly expressed in kidney and prostate, not founded in the liver, heart and pancreas. ARG2 has been implicated in the regulation of the arginine/ornithine concentrations in the cell. ARG2 may take part in the regulation of extra-urea cycle arginine metabolism and in down-regulation of nitric oxide synthesis. The extrahepatic arginase functions to regulate L-arginine bioavailability to NO synthase.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.