Human recombinant protein disulfide-isomerase A5 (from cells)
Catalog # PRSI91-965
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:Protein disulfide-isomerase A5
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:28.8 kD
- Sequence:Ser22-Leu262
- Formulation:Supplied as a 0.2 µm filtered solution of PBS,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Protein disulfide-isomerase A5 is a 519 amino acids protein that belongs to the protein disulfide isomerase family.It contains 3 thioredoxin domains.It can catalyse the rearrangement of -S-S- bonds in proteins.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.