Order Entry
Denmark
ContactUsLinkComponent
Human recombinant protein disulfide-isomerase A3 (from cells)
Human recombinant protein disulfide-isomerase A3 (from cells)
Catalog # PRSI91-869
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant protein disulfide-isomerase A3 (from cells)
Catalog # PRSI91-869
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Protein disulfide-isomerase A3
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    55.3 kD
  • Sequence:
    Ser25-Leu505
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

PDIA3 protein is also known as Protein disulfide-isomerase A3. It is a protein that in humans is encoded by the PDIA3 gene.PDIA3 is an enzyme that belongs to the endoplasmic reticulum and interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesised glycoproteins. PDIA3 interacts with thiazide-sensitive sodium-chloride cotransporter in the kidney and is induced by glucose deprivation. PDIA3 is part of the major histocompatibility complex (MHC) class I peptide-loading complex (TAP1), which is important for formation of the final antigen conformation and export from the endoplasmic reticulum to the cell surface.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.