Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Carboxypeptidase M (from Cells)
Human recombinant Carboxypeptidase M (from Cells)
Catalog # PRSI91-841
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Carboxypeptidase M (from Cells)
Catalog # PRSI91-841
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Carboxypeptidase M
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    47.3 kD
  • Sequence:
    Leu18-His422
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM ZnCl₂,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

These enzymes remove C-terminal amino acids from peptides and proteins and exert roles in the physiological processes of blood coagulation/fibrinolysis, inflammation, food digestion and pro-hormone and neuropeptide processing. CPM is widely distributed in a variety of tissues and cells. CPM is involved in peptide metabolism on both the cell surface and in extracellular fluids. CPM functions not only as a protease but also as a binding partner in cell-surface protein-protein interactions.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.