Order Entry
Denmark
ContactUsLinkComponent
Human recombinant Galactokinase 2 (from cells)
Human recombinant Galactokinase 2 (from cells)
Catalog # PRSI91-794
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Galactokinase 2 (from cells)
Catalog # PRSI91-794
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Galactokinase 2
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    51.4 kD
  • Sequence:
    Met1-Ala458
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

GALK2 acts as a galactokinase when galactose is present at high concentrations. GALK2 may be involved in a salvage pathway for the reutilisation of free GalNAc derived from the degradation of complex carbohydrates. GALK2 has been reported to participate in pathways , such as Amino sugar and nucleotide sugar metabolism, Galactose metabolism and Metabolic pathways.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.